missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, Anhydrous, powder, ≥97%, Honeywell Fluka™
Anhydrous, powder,≥97%
88.22 € - 441.00 €
Chemical Identifiers
CAS | 10043-52-4 |
---|---|
Molecular Formula | CaCl2 |
Molecular Weight (g/mol) | 110.98 |
MDL Number | MFCD00010903 |
InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
Synonym | calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal |
PubChem CID | 5284359 |
ChEBI | CHEBI:3312 |
SMILES | [Cl-].[Cl-].[Ca++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627760
|
Honeywell-Fluka
C4901-100G |
100 g | Poly Bottle |
88.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15657760
|
Honeywell-Fluka
C4901-500G |
500 g | Poly Bottle |
215.67 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15637760
|
Honeywell-Fluka
C4901-1KG |
1 kg | Poly bottle |
339.23 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15647760
|
Honeywell-Fluka
C4901-2.5KG |
2.5 kg | Poly Bottle |
385.61 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15667760
|
Honeywell-Fluka
C4901-5KG |
5 kg | Poly Bottle | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10043-52-4 | |
110.98 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
5284359 | |
[Cl-].[Cl-].[Ca++] |
CaCl2 | |
MFCD00010903 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
CHEBI:3312 |
Specifications
1,900°C | |
10043-52-4 | |
Powder | |
CaCl2 | |
NONH for all modes of transport | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium;dichloride | |
5284359 | |
110.98 | |
0.01 mmHg (20°C) | |
Calcium chloride, Anhydrous, powder, ≥97% |
772°C (literature) | |
100.0000% | |
CaCl2 | |
MFCD00010903 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
[Cl-].[Cl-].[Ca++] | |
110.98 | |
CHEBI:3312 | |
>97% | |
2.150 g/cm3 |
Safety and Handling
P305 + P351 + P338
missing translation for 'shelfLife' : 1620 days from date of manufacture
missing translation for 'einecsNumber' : 233-140-8
missing translation for 'rtecsNumber' : EV9800000
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcium chloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : calcium chloride