missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, ACS reagent, desiccant, Thermo Scientific Chemicals
51.26 € - 162.81 €
Chemical Identifiers
CAS | 10043-52-4 |
---|---|
Molecular Formula | CaCl2 |
Molecular Weight (g/mol) | 110.98 |
InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
Synonym | calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal |
PubChem CID | 5284359 |
ChEBI | CHEBI:3312 |
SMILES | [Cl-].[Cl-].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12664987
|
Thermo Scientific Acros
206791000 |
100 g |
51.26 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12614957
|
Thermo Scientific Acros
206795000 |
500 g |
162.81 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-52-4 | |
110.98 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
CHEBI:3312 |
CaCl2 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
5284359 | |
[Cl-].[Cl-].[Ca++] |
Specifications
>1600.0°C | |
10043-52-4 | |
CaCl2 | |
15,1661 | |
Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol | |
[Cl-].[Cl-].[Ca++] | |
110.98 | |
CHEBI:3312 | |
ACS Reagent | |
Calcium chloride |
782.0°C | |
CaCl2 | |
01,104; 04,368 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium;dichloride | |
5284359 | |
110.99 | |
Plastic bottle |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 233-140-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcium chloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Calcium chloride
RUO – Research Use Only