missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals

84.72 € - 145.73 €

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15455199
View Documents
Thermo Scientific Alfa Aesar
J62905.AP
500 mL
84.72 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15465199
View Documents
Thermo Scientific Alfa Aesar
J62905.K2
1 L
145.73 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10043-52-4
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
[Cl-].[Cl-].[Ca++]
CaCl2
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312

Specifications

10043-52-4
CaCl2
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312
100mM aq. soln.
Liquid
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
5284359
110.99
Calcium chloride, sterile
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.