missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, Light Powder, U.S.P. - F.C.C., J.T. Baker™
348.00 € - 1060.00 €
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15567894
|
Avantor J.T.Baker
1301-01 |
500 g | Poly Bottle |
348.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15577894
|
Avantor J.T.Baker
1301-05 |
2.5 kg | Poly Pail |
1060.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
471-34-1 | |
CCaO3 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
FCC/USP |
Powder | |
MFCD00010906 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
Calcium Carbonate |