missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate, 99+%, for biochemistry, Thermo Scientific Chemicals
39.50 € - 132.00 €
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12654977
|
Thermo Scientific Acros
192721000 |
100 g | Plastic bottle |
39.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12664977
|
Thermo Scientific Acros
192725000 |
500 g | Plastic bottle |
132.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
825.0°C | |
99% min. (Complexometry) | |
CaCO3 | |
01,103; 02,57; 04,67; 05,89 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
Biochemical |
471-34-1 | |
CCaO3 | |
MFCD00010906 | |
15,1659 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted acids | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
≥99% | |
Calcium carbonate |
Safety and Handling
missing translation for 'einecsNumber' : 207-439-9
RUO – Research Use Only