Learn More
Calcium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
75.30 € - 1739.00 €
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324728
|
Thermo Scientific Alfa Aesar
043073.06 |
5 g |
75.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354728
|
Thermo Scientific Alfa Aesar
043073.14 |
25 g |
259.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344728
|
Thermo Scientific Alfa Aesar
043073.22 |
100 g |
794.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334728
|
Thermo Scientific Alfa Aesar
043073.36 |
500 g |
1739.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
800°C (decomposition) | |
White | |
99.997% (Metal basis) | |
MFCD00010906 | |
Soluble in acids and insoluble in water. | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
1.6583 | |
2.93 |
471-34-1 | |
Powder | |
CCaO3 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
100.09 | |
White powder | |
Calcium carbonate |
Safety and Handling
missing translation for 'einecsNumber' : 207-439-9
missing translation for 'rtecsNumber' : FF9335000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only