missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbide, 72-82%, Thermo Scientific Chemicals

Product Code. 10219103
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-20-7
Molecular Formula C2Ca
Molecular Weight (g/mol) 64.1
MDL Number MFCD00010905
Synonym calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate

Specifications

Boiling Point 2300.0°C
Melting Point 2160.0°C
Color Gray
Physical Form Lump
Quantity 2.5 kg
Assay Percent Range 12 to 23% (CaO) (Complexometry)
Linear Formula CaC2
Merck Index 15, 1658
Solubility Information Solubility in water: hydrolyses
Formula Weight 64.1
Percent Purity 72 to 82%
Identification Positive
Packaging Glass bottle
Chemical Name or Material Calcium carbide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.