Learn More
Calcium bromide, ultra dry , 99.978% (metals basis), Thermo Scientific Chemicals
59.80 € - 158.79 €
Chemical Identifiers
CAS | 7789-41-5 |
---|---|
Molecular Formula | Br2Ca |
Molecular Weight (g/mol) | 199.886 |
MDL Number | MFCD00010902 |
InChI Key | WGEFECGEFUFIQW-UHFFFAOYSA-L |
Synonym | calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g |
PubChem CID | 5360316 |
ChEBI | CHEBI:31338 |
IUPAC Name | calcium;dibromide |
SMILES | [Ca+2].[Br-].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11373857
|
Thermo Scientific Alfa Aesar
035684.03 |
1 g |
59.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363857
|
Thermo Scientific Alfa Aesar
035684.06 |
5 g |
158.79 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaBr2 is mainly used as a dense aqueous solution for drilling fluid. It is also used in neuroses medication, freezing mixtures, food preservatives, photography and fire retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7789-41-5 | |
199.886 | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
5360316 | |
calcium;dibromide |
Br2Ca | |
MFCD00010902 | |
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g | |
CHEBI:31338 | |
[Ca+2].[Br-].[Br-] |
Specifications
806°C to 812°C | |
7789-41-5 | |
Beads | |
CaBr2 | |
14,1654 | |
It is soluble in water,alcohol and acetone. | |
[Ca+2].[Br-].[Br-] | |
199.886 | |
CHEBI:31338 | |
99.978% | |
Ampule | |
3.353 g/mL | |
Calcium bromide, Ultra dry |
730°C | |
White | |
Br2Ca | |
MFCD00010902 | |
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
calcium;dibromide | |
5360316 | |
199.9 | |
Odorless | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318
missing translation for 'einecsNumber' : 232-164-6
missing translation for 'rtecsNumber' : EV9328000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.