Learn More
Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals
77.70 € - 520.00 €
Chemical Identifiers
CAS | 1306-25-8 |
---|---|
Molecular Formula | CdTe |
Molecular Weight (g/mol) | 240.01 |
MDL Number | MFCD00015998 |
InChI Key | RPPBZEBXAAZZJH-UHFFFAOYSA-N |
Synonym | cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis |
PubChem CID | 91501 |
IUPAC Name | tellanylidenecadmium |
SMILES | [Cd]=[Te] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377917
|
Thermo Scientific Alfa Aesar
014365.06 |
5 g |
77.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357917
|
Thermo Scientific Alfa Aesar
014365.14 |
25 g |
179.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367917
|
Thermo Scientific Alfa Aesar
014365.22 |
100 g |
520.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Cadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles. .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
1306-25-8 | |
240.01 | |
RPPBZEBXAAZZJH-UHFFFAOYSA-N | |
91501 | |
[Cd]=[Te] |
CdTe | |
MFCD00015998 | |
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis | |
tellanylidenecadmium |
Spezifikation
1041°C | |
Black | |
99.999% (Metals basis) | |
MFCD00015998 | |
Insoluble in water. | |
[Cd]=[Te] | |
240.01 | |
240 | |
6.2 |
1306-25-8 | |
Polycrystalline Lump | |
CdTe | |
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis | |
RPPBZEBXAAZZJH-UHFFFAOYSA-N | |
tellanylidenecadmium | |
91501 | |
Odorless | |
Cadmium telluride |
Sicherheit und Handhabung
P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P501c
H332-H350-H372
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 215-149-9
missing translation for 'rtecsNumber' : EV3330000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only