Learn More
Cadmium sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
178.89 € - 691.44 €
Chemical Identifiers
CAS | 1306-23-6 |
---|---|
Molecular Formula | CdS |
Molecular Weight (g/mol) | 144.47 |
MDL Number | MFCD00010922 |
InChI Key | FRLJSGOEGLARCA-UHFFFAOYSA-N |
Synonym | cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow |
PubChem CID | 14783 |
ChEBI | CHEBI:50833 |
IUPAC Name | cadmium(2+) sulfanediide |
SMILES | [S--].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324457
|
Thermo Scientific Alfa Aesar
038342.18 |
50 g |
178.89 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334457
|
Thermo Scientific Alfa Aesar
038342.30 |
250 g |
691.44 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The main application of cadmium sulfide is as a pigment. Cadmium sulfide is also used in the production of solar cells. Used as light dependent resistors/photo resistors for light sensors. Used in MOCVD, sputtering and electrochemical deposition. In screen printing, it is used as a slurry containing dispersed cadmium sulfide. It is also used in dandruff shampoos.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-23-6 | |
144.47 | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
14783 | |
cadmium(2+) sulfanediide |
CdS | |
MFCD00010922 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
CHEBI:50833 | |
[S--].[Cd++] |
Specifications
980°C (sublimation) | |
Orange | |
CdS | |
UN3077 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
cadmium(2+) sulfanediide | |
14783 | |
144.46 | |
Odorless | |
(metals basis) |
1306-23-6 | |
Powder/Granules | |
MFCD00010922 | |
14,1628 | |
Soluble in acid,very slightly soluble in ammonium hydroxide. | |
[S--].[Cd++] | |
144.47 | |
CHEBI:50833 | |
99.999% | |
4.82 g/mL | |
Cadmium sulfide |
Safety and Handling
GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H350-H361fd-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-147-8
missing translation for 'rtecsNumber' : EV3150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.