Learn More
Cadmium selenide, 99.995% (metals basis), Thermo Scientific Chemicals
Vacuum deposition
64.70 € - 723.00 €
Chemical Identifiers
CAS | 1306-24-7 |
---|---|
Molecular Formula | CdSe |
Molecular Weight (g/mol) | 191.39 |
MDL Number | MFCD00010917 |
InChI Key | AQCDIIAORKRFCD-UHFFFAOYSA-N |
Synonym | cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene |
PubChem CID | 14784 |
ChEBI | CHEBI:50834 |
IUPAC Name | selanylidenecadmium |
SMILES | [Se]=[Cd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11383408
|
Thermo Scientific Alfa Aesar
036323.06 |
5 g |
64.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303418
|
Thermo Scientific Alfa Aesar
036323.14 |
25 g |
235.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393408
|
Thermo Scientific Alfa Aesar
036323.22 |
100 g |
723.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It ia a photoconductive and semiconductor material used in a cell where a fast response time and high sensitivity to longer wavelengths of light is required; also used as a red pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-24-7 | |
191.39 | |
AQCDIIAORKRFCD-UHFFFAOYSA-N | |
14784 | |
selanylidenecadmium |
CdSe | |
MFCD00010917 | |
cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene | |
CHEBI:50834 | |
[Se]=[Cd] |
Specifications
>1350°C | |
Black to Brown-Red | |
CdSe | |
UN3077 | |
cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene | |
AQCDIIAORKRFCD-UHFFFAOYSA-N | |
selanylidenecadmium | |
14784 | |
191.36 | |
Odorless | |
(metals basis) |
1306-24-7 | |
Sintered Lump | |
MFCD00010917 | |
14,1626 | |
Insoluble in water but soluble in acids. | |
[Se]=[Cd] | |
191.39 | |
CHEBI:50834 | |
99.995% | |
5.81 g/mL | |
Cadmium selenide |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350-H373
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-148-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only