Learn More
Cadmium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cadmium powder, 99.5%, Cd, CAS Number-7440-43-9, ccris 112, unii-00bh33gngh, compounds, cadmium, elemental, cadmium and compounds, 00bh33gngh, kadmium, kadmium german, colloidal, hsdb 282, 500g, (metals basis), CHEBI:22977, Silver-White
30.00 € - 917.00 €
Chemical Identifiers
CAS | 7440-43-9 |
---|---|
Molecular Formula | Cd |
Molecular Weight (g/mol) | 112.41 |
MDL Number | MFCD00010914 |
InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
Synonym | kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds |
PubChem CID | 23973 |
ChEBI | CHEBI:22977 |
IUPAC Name | cadmium |
SMILES | [Cd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341988
|
Thermo Scientific Alfa Aesar
011891.09 |
10 g |
30.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361988
|
Thermo Scientific Alfa Aesar
011891.22 |
100 g |
218.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351988
|
Thermo Scientific Alfa Aesar
011891.36 |
500 g |
917.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-43-9 | |
112.41 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
23973 | |
cadmium |
Cd | |
MFCD00010914 | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
CHEBI:22977 | |
[Cd] |
Specifications
Silver-White | |
Cd | |
UN3179 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
cadmium | |
23973 | |
99.5% | |
Solid | |
Cadmium powder |
7440-43-9 | |
MFCD00010914 | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
[Cd] | |
112.41 | |
CHEBI:22977 | |
−325 Mesh | |
(metals basis) |
Safety and Handling
GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P370+P378q-P501c
H228-H330-H341-H350-H361fd-H372
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 231-152-8
missing translation for 'rtecsNumber' : EU9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only