Learn More
Cadmium phosphate (ortho), 99%, Thermo Scientific Chemicals
51.90 € - 692.00 €
Chemical Identifiers
CAS | 13847-17-1 |
---|---|
Molecular Formula | Cd3O8P2 |
Molecular Weight (g/mol) | 527.182 |
MDL Number | MFCD00049414 |
InChI Key | NRGIRRZWCDKDMV-UHFFFAOYSA-H |
Synonym | cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate |
PubChem CID | 26060 |
IUPAC Name | cadmium(2+);diphosphate |
SMILES | [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319347
|
Thermo Scientific Alfa Aesar
040201.09 |
10 g | |||||||
11309347
|
Thermo Scientific Alfa Aesar
040201.18 |
50 g |
183.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399337
|
Thermo Scientific Alfa Aesar
040201.30 |
250 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13847-17-1 | |
527.182 | |
NRGIRRZWCDKDMV-UHFFFAOYSA-H | |
26060 | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2] |
Cd3O8P2 | |
MFCD00049414 | |
cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate | |
cadmium(2+);diphosphate |
Specifications
∼1500°C | |
Powder | |
MFCD00049414 | |
cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate | |
NRGIRRZWCDKDMV-UHFFFAOYSA-H | |
cadmium(2+);diphosphate | |
26060 | |
99% |
13847-17-1 | |
Cd3O8P2 | |
UN2570 | |
Insoluble in water | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2] | |
527.182 | |
527.14 | |
Cadmium phosphate (ortho) |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
missing translation for 'tsca' : TSCA
RUO – Research Use Only