Learn More
Cadmium molybdenum oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Electronic and optical applications
131.00 € - 543.71 €
Chemical Identifiers
CAS | 13972-68-4 |
---|---|
Molecular Formula | CdMoO4 |
Molecular Weight (g/mol) | 272.36 |
MDL Number | MFCD00015997 |
InChI Key | SQHWLBOPUKZGNS-UHFFFAOYSA-N |
Synonym | cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum |
PubChem CID | 22420716 |
IUPAC Name | cadmium(2+);dioxido(dioxo)molybdenum |
SMILES | [O--].[O--].[O--].[O--].[Mo].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329347
|
Thermo Scientific Alfa Aesar
040202.18 |
50 g | N/A | ||||||
11339347
|
Thermo Scientific Alfa Aesar
040202.30 |
250 g |
543.71 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Electronic and optical applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13972-68-4 | |
272.36 | |
SQHWLBOPUKZGNS-UHFFFAOYSA-N | |
22420716 | |
[O--].[O--].[O--].[O--].[Mo].[Cd++] |
CdMoO4 | |
MFCD00015997 | |
cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum | |
cadmium(2+);dioxido(dioxo)molybdenum |
Specifications
∼1250°C | |
Powder | |
MFCD00015997 | |
cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum | |
SQHWLBOPUKZGNS-UHFFFAOYSA-N | |
cadmium(2+);dioxido(dioxo)molybdenum | |
22420716 | |
99.5% | |
(metals basis) |
13972-68-4 | |
CdMoO4 | |
UN2570 | |
Slightly soluble in water; Soluble in acid | |
[O--].[O--].[O--].[O--].[Mo].[Cd++] | |
272.36 | |
272.35 | |
Odorless | |
Cadmium molybdenum oxide |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
missing translation for 'tsca' : TSCA
RUO – Research Use Only