Learn More
Cadmium iodide, 98%, Thermo Scientific Chemicals
33.10 € - 98.30 €
Chemical Identifiers
CAS | 7790-80-9 |
---|---|
Molecular Formula | CdI2 |
Molecular Weight (g/mol) | 366.223 |
MDL Number | MFCD00010920 |
InChI Key | OKIIEJOIXGHUKX-UHFFFAOYSA-L |
Synonym | cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g |
PubChem CID | 24635 |
IUPAC Name | cadmium(2+);diiodide |
SMILES | [Cd+2].[I-].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429226
|
Thermo Scientific Alfa Aesar
A18079.14 |
25 g |
33.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439226
|
Thermo Scientific Alfa Aesar
A18079.22 |
100 g |
98.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.
Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
7790-80-9 | |
366.223 | |
OKIIEJOIXGHUKX-UHFFFAOYSA-L | |
24635 | |
[Cd+2].[I-].[I-] |
CdI2 | |
MFCD00010920 | |
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g | |
cadmium(2+);diiodide |
Specifications
787°C | |
7790-80-9 | |
MFCD00010920 | |
14,1621 | |
Very soluble in water,slightly soluble in acetone,ethanol and diethyl ether. | |
[Cd+2].[I-].[I-] | |
366.223 | |
366.21 | |
Odorless | |
5.67 |
387°C to 389°C | |
CdI2 | |
UN2570 | |
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g | |
OKIIEJOIXGHUKX-UHFFFAOYSA-L | |
cadmium(2+);diiodide | |
24635 | |
98% | |
Hygroscopic; Light sensitive | |
Cadmium iodide |
Safety and Handling
GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H351-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
missing translation for 'einecsNumber' : 232-223-6
missing translation for 'rtecsNumber' : EV1290000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only