missing translation for 'onlineSavingsMsg'
Learn More

Cadmium, course powder, 1-4mm for reductors, ≥99% (metals basis), Thermo Scientific™

Product Code. 15457495
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

UN Number UN2570
Quantity 100 g
Percent Purity ≥99%
Form Coarse Powder
Assay (metals basis)
Chemical Name or Material Cadmium, 1 to 4mm for reductors
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'