missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

110.55 € - 374.87 €

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386928
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013667.06
5 g
110.55 €
5g
Please to purchase this item. Need a web account? Register with us today!
11376928
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013667.14
25 g
374.87 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10108-64-2
183.31
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
CHEBI:35456
[Cl-].[Cl-].[Cd++]

Specifications

960°C
10108-64-2
CdCl2
UN2570
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.32
Odorless
Hygroscopic
(metals basis)
568°C
Beads
MFCD00010916
14,1617
Soluble in water and acetone.
[Cl-].[Cl-].[Cd++]
183.31
CHEBI:35456
99.998%
Ampule
4.047 g/mL
Cadmium chloride, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H335-H340-H350-H360FD-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

missing translation for 'einecsNumber' : 233-296-7

missing translation for 'rtecsNumber' : EV0175000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only