missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals

85.53 € - 315.57 €

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
IUPAC Name cadmium(2+) carbonate
SMILES [Cd++].[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341938
SDS View Documents
Thermo Scientific Alfa Aesar
011864.14
25 g
85.53 €
25g
Please to purchase this item. Need a web account? Register with us today!
11331938
SDS View Documents
Thermo Scientific Alfa Aesar
011864.22
100 g
315.57 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium carbonate, has been used as intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer,and, raw material for production of cadmium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-78-0
172.42
GKDXQAKPHKQZSC-UHFFFAOYSA-L
10564
[Cd++].[O-]C([O-])=O
CCdO3
MFCD00010918
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
cadmium(2+) carbonate

Specifications

500°C (decomposition)
White
99.99%
MFCD00010918
It is insoluble in water and organic solvents,slightly soluble in aqueous acid,potassium cyanide,ammonium.
[Cd++].[O-]C([O-])=O
172.42
172.42
White powder/lumps
Cadmium carbonate
513-78-0
Lump/Powder
CCdO3
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+) carbonate
10564
Odorless
4.26 g/mL
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H302+H312-H330-H340-H350-H361-H372

missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 208-168-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.