missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 15289886
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-42-6
Molecular Formula Br2Cd
Molecular Weight (g/mol) 272.22
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 963.0°C
Melting Point 566.0°C
Quantity 25 g
Assay Percent Range 97.5% min
Merck Index 15,162
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, moderatly soluble in acetone, slightly soluble in ether
Formula Weight 272.22
Percent Purity 98%
Grade Pure
Chemical Name or Material Cadmium bromide
compliance-icons compliance-icons
Product Identifier
  • Cadmium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'