Learn More
Cacodylic acid, 98%, Thermo Scientific Chemicals
35.98 € - 661.29 €
Chemical Identifiers
CAS | 75-60-5 |
---|---|
Molecular Formula | C2H7AsO2 |
Molecular Weight (g/mol) | 138.00 |
MDL Number | MFCD00002095 |
InChI Key | OGGXGZAMXPVRFZ-UHFFFAOYSA-N |
Synonym | cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 |
PubChem CID | 2513 |
ChEBI | CHEBI:48765 |
IUPAC Name | dimethylarsinic acid |
SMILES | C[As](C)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10770785
|
Thermo Scientific Acros
318150050 |
5 g | Glass bottle |
35.98 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10711551
|
Thermo Scientific Acros
318150100 |
10 g | Glass bottle |
50.15 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15739128
|
Thermo Scientific Acros
318150500 |
50 g | Glass bottle |
171.86 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10701355
|
Thermo Scientific Acros
318152500 |
250 g | Glass bottle |
661.29 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-60-5 | |
138.00 | |
OGGXGZAMXPVRFZ-UHFFFAOYSA-N | |
2513 | |
dimethylarsinic acid |
C2H7AsO2 | |
MFCD00002095 | |
cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 | |
CHEBI:48765 | |
C[As](C)(O)=O |
Specifications
75-60-5 | |
White | |
C2H7AsO2 | |
MFCD00002095 | |
cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 | |
C[As](C)(O)=O | |
138.00 | |
CHEBI:48765 | |
≥97.5% | |
Crystalline Powder |
195°C to 198°C | |
98% | |
(CH3)2As(O)OH | |
15, 1609 | |
OGGXGZAMXPVRFZ-UHFFFAOYSA-N | |
dimethylarsinic acid | |
2513 | |
138.01 | |
Authentic | |
Cacodylic acid |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.