Learn More
YSI™ Cables and DO/Temp/Conductivity Probes for 556 Multi-Probe System
Handheld multiprobe meter
Brand:  YSI™ 55634
 View more versions of this product
Product Code. 11930105
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
DO/Temperature/Conductivity Probe and Cable. Required to measure dissolved oxygen, temperature, conductivity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13 ft.
Temperature/conductivity sensor
YSI model 556 handheld multi probe meter
4 m
1 Year
Measures DO, temperature, conductivity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products