Learn More
Description
Semi Dry Blotting has the benefit of economic transfers due to very low buffer volumes, typically only a few mL per transfer. SD-10 and SD-20 Semi-Dry Blotters have a screw down lid, which secures the blot sandwich and allows complete control of pressure ensuring even transfer. The electrodes, comprising platinum coated anode and stainless steel cathode, exhibit virtually no corrosion for many years of trouble free use.
- Set of two cables: red (positive) and black (negative)
- Red lead inserts through the lid; black lead inserts through the side of the base
- Compatible with SD-10 Semi-Dry Blotter (10 x 10cm) and SD-20 Semi-Dry Blotter (20 x 20cm)
Specifications
Red/Black | |
SD-10 and SD-20 Semi-Dry Blotters | |
Positive and Negative leads |
Electrophoresis | |
Power Cables | |
2 Cables |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.