Learn More
CA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13198-2-AP-20UL
 View more versions of this product
Product Code. 16881294
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.
Specifications
Specifications
CA1
Polyclonal
Unconjugated
CA1
CA I, CA1, CAB, Car1, Carbonate dehydratase I, Carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase I
Rabbit
Antigen Affinity Chromatography
RUO
12346, 310218, 759
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00915, P13634
CA1, Car1
CA1 Fusion Protein Ag3932
20 μL
Primary
Rat, Mouse, Human