missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf24 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | C5orf24 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882795
|
Proteintech
24319-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872795
|
Proteintech
24319-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C5orf24 is a protein coding gene.Specifications
C5orf24 | |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q7Z6I8 | |
134553 | |
C5orf24 Fusion Protein Ag19534 | |
Primary | |
-20°C | |
C5orf24 |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C5orf24, UPF0461 protein C5orf24 | |
C5orf24 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title