missing translation for 'onlineSavingsMsg'
Learn More

Butyryl chloride, 99%, Thermo Scientific Chemicals

22.91 € - 139.70 €

Chemical Identifiers

CAS 141-75-3
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000752
InChI Key DVECBJCOGJRVPX-UHFFFAOYSA-N
Synonym butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
PubChem CID 8855
IUPAC Name butanoyl chloride
SMILES CCCC(Cl)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10701744
View Documents
Thermo Scientific
149210500
50 mL Glass bottle
22.91 €
50mL
Please to purchase this item. Need a web account? Register with us today!
10153720
View Documents
Thermo Scientific Acros
149212500
250 mL Glass bottle
47.54 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10245290
View Documents
Thermo Scientific Acros
149210010
1 L Glass bottle
139.70 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-75-3
106.55
DVECBJCOGJRVPX-UHFFFAOYSA-N
8855
CCCC(Cl)=O
C4H7ClO
MFCD00000752
butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
butanoyl chloride

Specifications

141-75-3
Yellow
102.0°C
Authentic
C4H7ClO
1.4112 to 1.4132
02, 274
1.018
Solubility in water: decomposes. Other solubilities: miscible with ether,decomposes in alcohol
CCCC(Cl)=O
106.55
106.55
Liquid
-89.0°C
1.0180g/mL
18°C
99%
CH3CH2CH2COCl
MFCD00000752
15, 1602
butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
DVECBJCOGJRVPX-UHFFFAOYSA-N
butanoyl chloride
8855
99%
Butyryl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 205-498-5

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.