missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 9.18, Reagecon™
BIOPHARMA

Product Code. 11701389 Shop All Reagecon Products
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a NIST/DIN certified pH buffer. Mettler Toledo™ Buffer Solution pH 9.18 comes in a 250mL bottle.

  • NIST/DIN certification
  • pH 9180
  • 250ML

Specifications

pH 9.18
Quantity 250 mL
compliance-icons
Product Identifier
  • SOLUTION TAMPON pH 9.180 a 25°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : boric acid, disodiumsalt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.