missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 8.00, BAKER ANALYZED, J.T. Baker™
Click to view available options
Quantity:
1 L
Description
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
Density | 1g/cm3 at 20°C |
Physical Form | Liquid |
Quantity | 1 L |
Formulation | Contains Potassium dihydrogen phosphate and Sodium hydroxide |
Solubility Information | Fully miscible in water |
Grade | BAKER ANALYZED™ |
Chemical Name or Material | Phosphate buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction