missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 7.413 (25°C), With fungicide, according to DIN 19266, Honeywell Fluka™
With fungicide, according to DIN 19266
Brand: Honeywell-Fluka 33597-6X1L
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer solution pH 7.413 (25°C) | |
100°C | |
≤100.0000%, ≤1.0000% | |
1.000 g/cm3 | |
Buffer |
With fungicide, according to DIN 19266 | |
≥90.0000% | |
7558-79-4, 7778-77-0 | |
6 x 1 L | |
Poly Bottle |