missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 4.01, METTLER TOLEDO™
The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the perfect product for regulated industries.
112.00 €
Specifications
Color | Light Red |
---|---|
Description | Certified buffer pH 4.01, 6 x 250mL |
pH | 4.01 |
Physical Form | Liquid |
Solution Type | pH Buffer |
Odor | None |
Packaging | 6x 250 mL |
Description
Specifications
Light Red | |
4.01 | |
pH Buffer | |
6x 250 mL |
Certified buffer pH 4.01, 6 x 250mL | |
Liquid | |
None |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title