Learn More
Buffer solution, pH 2.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
For Calibration of pH Meters
27.50 € - 80.40 €
Specifications
Chemical Name or Material | Buffer solution |
---|---|
CAS | 7447-40-7, 7647-01-0, 7732-18-5 |
Color | Colorless |
Physical Form | Liquid |
MDL Number | MFCD00146252 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480271
|
Thermo Scientific Alfa Aesar
040447.AP |
500 mL |
27.50 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358408
|
Thermo Scientific Alfa Aesar
040447.K2 |
1 L |
43.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348408
|
Thermo Scientific Alfa Aesar
040447.K7 |
4 L |
80.40 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Buffer Solution pH 2 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine Buffer Solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
MFCD00146252 | |
23 hPa (17mm Hg) at 20°C | |
Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6% |
7447-40-7, 7647-01-0, 7732-18-5 | |
Liquid | |
Yes | |
Soluble in water. |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.
P234-P390
H290
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only