missing translation for 'onlineSavingsMsg'
Learn More

Color Coded pH Buffer Solution Tested at 25°C, Reagecon™
BIOPHARMA

Product Code. 11494579 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle
HDPE Bottle GL45
Color:
Blue
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 25°C are certified and NIST traceable.

  • Colored pH Buffer Solutions
  • Tested at 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Color Blue
pH 10
Physical Form Liquid
Quantity 1 L
Packaging HDPE Bottle GL45
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.