missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10-11 [NH4Cl/NH4OH], BAKER ANALYZED™, J.T. Baker™
Click to view available options
1 L
Description
- Contains Ammonium Chloride / Ammonium Hydroxide
- This buffer is tracable to NIST Standard Reference Buffer
Specifications
Specifications
pH | 10 to 11 |
Quantity | 1 L |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction