missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 10.01 at 25 °C, Reagecon
Guaranteed stability throughout entire shelf life even after opening the bottle, NIST traceable
Brand: Reagecon 11001525
This item is not returnable.
View return policy
Description
- Tested by ISO 17025 accredited test method
Specifications
Buffer solution | |
Liquid | |
HDPE bottle GL45 |
10.01 ±0.01 | |
500 mL | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction