missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide, Honeywell Fluka™

Product Code. 15694040
Click to view available options
Quantity:
100 mL
500 mL
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Borax / sodium hydroxide

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 1330-43-4, 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

pH 10.0
Quantity 100 mL
UN Number NONH for all modes of transport
IUPAC Name amine hydrate
Chemical Name or Material Buffer solution
Hazard Statement
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'