missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solutions, WTW™

Product Code. 10078720
25 mL
Click to view available options
Quantity:
25 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Buffer solution that allows the evaluation possibility/data transfer to PC for documentation according to GLP with the respirometric OxiTop™ Control meter for BOD self check measurement.

  • Nitrification inhibitor
  • Accessory for BOD self check measurement OxiTop™ Control
  • Buffer solution that allows the evaluation possibility/data transfer to PC for documentation according to GLP with the respirometric OxiTop™ Control meter for BOD self check measurement
TRUSTED_SUSTAINABILITY

Specifications

Density 1g/cm3
pH 6.5
Physical Form Liquid
Quantity 25 mL
Packaging Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solutions, WTW™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.