Learn More
BSA Mouse, anti-Bovine, HRP, Clone: 2D11G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech HRP-66201-100UL
This item is not returnable.
View return policy
Description
BSA, also named as ALB and Bovine Serum albumin, is the main protein of plasma. It has a good binding capacity for water, Ca2+, Na+, K+, fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood.
Specifications
BSA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Liquid |
ELISA | |
2D11G3 | |
HRP | |
100 μL | |
Bovine |