Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2640283
 View more versions of this product
Product Code. 30090420
  • 86.15 € / Pack of 2
Explore more special offers
Description
Description
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
To Contain
Brown
Brown
Borosilicate Glass (3.3)
Polyethylene Stopper
2/Pack
± 0.15 mL
Autoclavable
No
A
NS 14/23
DIN EN ISO 1042
Polyethylene
150 mL
180 ± 5 mm
± 0.1 mL
14/23 NS Standard Ground
White enamel color
Reusable
Non-sterile
Round