missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brooker's merocyanine dye

203.01 € - 708.53 €

Chemical Identifiers

CAS 23302-83-2
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00010099
InChI Key DBOHWMPKJCJANT-UHFFFAOYSA-N
Synonym brooker's merocyanine, 4-2-1-methylpyridin-4 1h-ylidene ethylidene cyclohexa-2,5-dien-1-one, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-dien-1-one, 2,5-cyclohexadien-1-one,4-2-1-methyl-4 1h-pyridinylidene ethylidene, brooker/'s merocyanine, 1-methyl-4-4-oxylatostyryl pyridinium, 1-methyl-4-e-4-oxylatostyryl pyridinium, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-die, 4-2-1-methyl-4-pyridylidene ethylidene cyclohexa-2,5-dien-1-one
PubChem CID 258436
IUPAC Name 4-[2-(1-methylpyridin-4-ylidene)ethylidene]cyclohexa-2,5-dien-1-one
SMILES CN1C=CC(=CC=C2C=CC(=O)C=C2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15462779
SDS View Documents
Thermo Scientific Chemicals
H66517.MD
250 mg
203.01 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15452779
SDS View Documents
Thermo Scientific Chemicals
H66517.03
1 g
708.53 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Brooker's merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brooker′s merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Brooker's merocyanine dye

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.