Learn More
Bromotrimethylsilane, 97%, stab. with copper powder or silver wire, Thermo Scientific Chemicals
41.00 € - 531.00 €
Chemical Identifiers
CAS | 2857-97-8 |
---|---|
Molecular Formula | C3H9BrSi |
Molecular Weight (g/mol) | 153.094 |
MDL Number | MFCD00000048 |
InChI Key | IYYIVELXUANFED-UHFFFAOYSA-N |
Synonym | trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide |
PubChem CID | 76113 |
IUPAC Name | bromo(trimethyl)silane |
SMILES | C[Si](C)(C)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488236
|
Thermo Scientific Alfa Aesar
A15334.09 |
10 g |
41.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498236
|
Thermo Scientific Alfa Aesar
A15334.18 |
50 g |
130.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408246
|
Thermo Scientific Alfa Aesar
A15334.30 |
250 g |
531.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.
Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
Chemical Identifiers
2857-97-8 | |
153.094 | |
IYYIVELXUANFED-UHFFFAOYSA-N | |
76113 | |
C[Si](C)(C)Br |
C3H9BrSi | |
MFCD00000048 | |
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide | |
bromo(trimethyl)silane |
Specifications
78°C to 80°C | |
2857-97-8 | |
(CH3)3SiBr | |
UN2924 | |
−3°C (26°F) | |
Miscible with carbon tetrachloride,chloroform,dichloroethane,acetonitrile,toluene,dichloromethane and hexane. | |
C[Si](C)(C)Br | |
153.094 | |
153.09 | |
Acrid | |
Moisture sensitive | |
Bromotrimethylsilane, stabilized with copper powder or silver wire |
-44°C | |
C3H9BrSi | |
MFCD00000048 | |
1731135 | |
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide | |
IYYIVELXUANFED-UHFFFAOYSA-N | |
bromo(trimethyl)silane | |
76113 | |
97% | |
1.425 | |
1.183 |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 220-672-0
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only