Learn More
Bromotrichloromethane, 99%, Thermo Scientific Chemicals
37.80 € - 70.80 €
Chemical Identifiers
CAS | 75-62-7 |
---|---|
Molecular Formula | CBrCl3 |
Molecular Weight (g/mol) | 198.27 |
MDL Number | MFCD00000783 |
InChI Key | XNNQFQFUQLJSQT-UHFFFAOYSA-N |
Synonym | methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br |
PubChem CID | 6383 |
IUPAC Name | bromo(trichloro)methane |
SMILES | C(Cl)(Cl)(Cl)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10715751
|
Thermo Scientific Acros
107491000 |
100 mL | Glass bottle |
37.80 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10050550
|
Thermo Scientific Acros
107492500 |
250 mL | Glass bottle |
70.80 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
75-62-7 | |
198.27 | |
XNNQFQFUQLJSQT-UHFFFAOYSA-N | |
6383 | |
C(Cl)(Cl)(Cl)Br |
CBrCl3 | |
MFCD00000783 | |
methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br | |
bromo(trichloro)methane |
Specifications
75-62-7 | |
Colorless to Yellow | |
105.0°C | |
98.5% min. (GC) | |
BrCCl3 | |
MFCD00000783 | |
01,80; 02,42; 07,40 | |
methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br | |
XNNQFQFUQLJSQT-UHFFFAOYSA-N | |
bromo(trichloro)methane | |
6383 | |
99% | |
Bromotrichloromethane, 99% |
-6.0°C | |
1.9970g/mL | |
Authentic | |
CBrCl3 | |
1.5055 to 1.5075 | |
01, 67 | |
1.997 | |
Solubility in water: insoluble. Other solubilities: miscible with many organic liquids, soluble in ethanol and ether | |
C(Cl)(Cl)(Cl)Br | |
198.27 | |
198.27 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-886-0
missing translation for 'rtecsNumber' : PA5400000
missing translation for 'tsca' : TSCA
- Bromotrichloromethane
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only