Learn More
(Bromomethyl)cyclobutane, 99%, Thermo Scientific Chemicals
57.08 € - 721.00 €
Chemical Identifiers
CAS | 17247-58-4 |
---|---|
Molecular Formula | C5H9Br |
Molecular Weight (g/mol) | 149.03 |
MDL Number | MFCD00040960 |
InChI Key | FLHFTXCMKFVKRP-UHFFFAOYSA-N |
Synonym | bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan |
PubChem CID | 2734658 |
IUPAC Name | bromomethylcyclobutane |
SMILES | C1CC(C1)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10223052
|
Thermo Scientific Acros
367640050 |
5 mL | Glass bottle |
57.08 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10384642
|
Thermo Scientific Acros
367640250 |
25 mL | Glass bottle |
194.97 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10544464
|
Thermo Scientific Acros
367641000 |
100 mL | Glass bottle |
721.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17247-58-4 | |
149.03 | |
FLHFTXCMKFVKRP-UHFFFAOYSA-N | |
2734658 | |
C1CC(C1)CBr |
C5H9Br | |
MFCD00040960 | |
bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan | |
bromomethylcyclobutane |
Specifications
17247-58-4 | |
100.0 | |
1.3390g/mL | |
27°C | |
98.5% min. (GC) | |
C4H7CH2Br | |
MFCD00040960 | |
1.339 | |
Solubility in water: .. Other solubilities: soluble in alcohols | |
C1CC(C1)CBr | |
149.03 | |
149.03 | |
Liquid |
98.5 | |
Colorless | |
145.0°C to 148.0°C | |
Authentic | |
C5H9Br | |
1.4790 to 1.4830 | |
05, 22 | |
bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan | |
FLHFTXCMKFVKRP-UHFFFAOYSA-N | |
bromomethylcyclobutane | |
2734658 | |
99% | |
(Bromomethyl)cyclobutane, 99% |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- (Bromomethyl)cyclobutane
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only