missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl acetate, 95%, Thermo Scientific Chemicals

291.00 € - 410.00 €

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000170
InChI Key NHYXMAKLBXBVEO-UHFFFAOYSA-N
Synonym bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
PubChem CID 68536
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403298
View Documents
Thermo Scientific Alfa Aesar
H56755.06
5 g
291.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
15413298
View Documents
Thermo Scientific Alfa Aesar
H56755.09
10 g
410.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

Solubility
Miscible with chloroform.

Notes
Store in cool place. Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

590-97-6
152.98
NHYXMAKLBXBVEO-UHFFFAOYSA-N
68536
CC(=O)OCBr
C3H5BrO2
MFCD00000170
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
bromomethyl acetate

Specifications

590-97-6
130°C to 133°C
C3H5BrO2
MFCD00000170
Moisture sensitive
Miscible with chloroform.
CC(=O)OCBr
152.98
152.98
Bromomethyl acetate
1.56
57°C (135°F)
CH3CO2CH2Br
UN3272
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
NHYXMAKLBXBVEO-UHFFFAOYSA-N
bromomethyl acetate
68536
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

missing translation for 'einecsNumber' : 209-696-2

missing translation for 'rtecsNumber' : AF6300200

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only