missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid N-hydroxysuccinimide ester, 90%, Thermo Scientific Chemicals

Product Code. 10594954
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42014-51-7
Molecular Formula C6H6BrNO4
Molecular Weight (g/mol) 236.02
MDL Number MFCD00058571
InChI Key NKUZQMZWTZAPSN-UHFFFAOYSA-N
Synonym n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
PubChem CID 3565210
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
SMILES C1CC(=O)N(C1=O)OC(=O)CBr

Specifications

Color White
Elemental Analysis 88% min.
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 1 g
Formula Weight 236.02
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Bromoacetic acid N-hydroxysuccinimide ester
compliance-icons
Product Identifier
  • Bromoacetic acid N-hydroxysuccinimide ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.