Learn More
Bromine liquid, ACS, 99.5%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 0.005%, Cl 0.05%, I 0.001%, Sulfur compounds (as S) 0.001%, Heavy Metals (as Pb) 2ppm, Ni 5ppm
26.80 € - 858.00 €
Chemical Identifiers
CAS | 7726-95-6 |
---|---|
Molecular Formula | Br |
Molecular Weight (g/mol) | 79.91 |
MDL Number | MFCD00010896 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-M |
Synonym | bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish |
PubChem CID | 24408 |
ChEBI | CHEBI:29224 |
SMILES | [Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15451317
|
Thermo Scientific Alfa Aesar
033220.09 |
10 g | N/A | ||||||
15461317
|
Thermo Scientific Alfa Aesar
033220.22 |
100 g | N/A | ||||||
15471317
|
Thermo Scientific Alfa Aesar
033220.36 |
500 g | N/A | ||||||
15481317
|
Thermo Scientific Alfa Aesar
033220.61 |
5 x 500 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7726-95-6 | |
79.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
24408 | |
[Br-] |
Br | |
MFCD00010896 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CHEBI:29224 |
Specifications
7726-95-6 | |
Liquid | |
Br | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
99.5% | |
Unpleasant | |
230 hPa (173mm Hg) at 20°C | |
Bromine liquid |
Brown to Red | |
99.5% | |
MFCD00010896 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
bromide | |
24408 | |
159.81 | |
ACS | |
Red-brown fuming liquid | |
3.12g/cm3 at 20°C |
Safety and Handling
P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H330
missing translation for 'dotInformation' : DOT Class: 8; Packing Group: I
missing translation for 'einecsNumber' : 231-778-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.