Learn More
Bromine, 99.6%, for analysis, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7726-95-6 |
---|---|
Molecular Formula | Br |
Molecular Weight (g/mol) | 79.91 |
MDL Number | MFCD00010896 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-M |
Synonym | bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish |
PubChem CID | 24408 |
ChEBI | CHEBI:29224 |
SMILES | [Br-] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent for analysisChemical Identifiers
7726-95-6 | |
79.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
24408 | |
[Br-] |
Br | |
MFCD00010896 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CHEBI:29224 |
Specifications
58.7°C | |
7726-95-6 | |
Br | |
MFCD00010896 | |
15, 1401 | |
Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2 | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
99.6% | |
Plastic Coated Glass Bottle | |
3.1110g/mL |
-7.2°C | |
99.5% min. (Iodometry) | |
Br2 | |
03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
molecular bromine | |
24408 | |
159.82 | |
Analytical | |
3.111 | |
Bromine, p.a. |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-778-1
missing translation for 'rtecsNumber' : EF9100000
missing translation for 'tsca' : TSCA
- Bromine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
RUO – Research Use Only