missing translation for 'onlineSavingsMsg'
Learn More

Brix Standard, Reagecon™
BIOPHARMA

Stabilized to have an extended 12-month shelf-life. Reagecon™ Brix Standard is a range of ready-to-use sucrose refractive index standards in a convenient dropper bottle for use with all types of refractometers. Test results accredited to ISO 17025 and manufactured in accordance with ISO Guide 34.

75.30 € - 106.00 €

Chemical Identifiers

CAS 57-50-1, 7732-18-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym Sucrose solution
PubChem CID 5988
ChEBI CHEBI:17992
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View More Specs

Products 4
Product Code Brand Concentration Price Quantity & Availability  
Product Code Brand Concentration Price Quantity & Availability  
13474016
BIOPHARMA
View Documents
Reagecon
BS10S
10% sucrose
75.30 €
15mL
Please to purchase this item. Need a web account? Register with us today!
13474026
BIOPHARMA
View Documents
Reagecon
BS30S
30% sucrose
75.30 €
15mL
Please to purchase this item. Need a web account? Register with us today!
13494016
BIOPHARMA
View Documents
Reagecon
BS05S
5% sucrose
75.30 €
15mL
Please to purchase this item. Need a web account? Register with us today!
13494036
BIOPHARMA
View Documents
Reagecon
BS60S
60% sucrose
106.00 €
15mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This range of sucrose standard solutions contains high purity raw ingredients, can be used with any brand of refractometer and is stabilized to have an extended shelf-life to provide excellent value for users that are not required to follow ICUMSA Guidelines.

  • Prepared gravimetrically on a weight/weight basis: both solute (sucrose) and solvent (water) are weighed on a balance calibrated using OIML traceable weights
  • Balance Certificate of Calibration is issued in accordance with ISO 17025 for calibration of nonautomatic weighing machines (INAB ref: 265C)
  • Brix value is verified using a high-performance calibrated, temperature-controlled refractometer using high-purity, ISO Guide 34-accredited Brix standards
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brix Standard, Reagecon™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.