Learn More
Macherey-Nagel™ Brilliant Yellow pH Paper
Brand:  Macherey-Nagel™ 90701
Product Code. 11326514
  • 26.05 € / Pack of 200
Explore more special offers
Specifications
Yellow to Red
Strip
1 Each
6.7 to 7.9
For determination of alkali concentration
pH Paper