Learn More
Thermo Scientific Chemicals Brilliant Green, pure, high purity biological stain
49.65 € - 160.80 €
Chemical Identifiers
CAS | 633-03-4 |
---|---|
Molecular Formula | C27H34N2O4S |
Molecular Weight (g/mol) | 482.639 |
MDL Number | MFCD00011880 |
InChI Key | NNBFNNNWANBMTI-UHFFFAOYSA-M |
Synonym | Basic Green 1, C.I. 42040, Ethyl Green |
PubChem CID | 12449 |
IUPAC Name | [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate |
SMILES | CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10195230
|
Thermo Scientific Chemicals
229601000 |
100 g | Glass bottle |
49.65 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10511794
|
Thermo Scientific Chemicals
229605000 |
500 g | Glass bottle |
160.80 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
633-03-4 | |
482.639 | |
NNBFNNNWANBMTI-UHFFFAOYSA-M | |
12449 | |
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-] |
C27H34N2O4S | |
MFCD00011880 | |
Basic Green 1, C.I. 42040, Ethyl Green | |
[4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate |
Specifications
1875 (at lambda min.) | |
633-03-4 | |
C27H34N2O4S | |
13, 746 | |
Basic Green 1, C.I. 42040, Ethyl Green | |
NNBFNNNWANBMTI-UHFFFAOYSA-M | |
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-] | |
482.639 | |
625 to 632nm (in 50:50 IMS:water) | |
10% max. (105°C) | |
Green | |
Brilliant Green, High purity biological stain |
180.0°C | |
1 to 1.2 | |
MFCD00011880 | |
15, 1380 | |
Solubility in water: 100g/L water (25°C). Other solubilities: soluble in alcohol | |
Authentic | |
[4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate | |
12449 | |
482.64 | |
Pure | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Brilliant Green, high purity biological stain
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.