Learn More
BRAND™ Replacement Dispenser Cartridges
Replacement Cartridges for seripettor™ and seripettor™ pro, and QuickSip bottletop dispensers
36.35 € - 115.00 €
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
10065460
|
BRAND™
704502 |
10 mL | Non-sterile |
36.35 €
Pack of 3 |
In Stock
Log in to see stock. |
||||
10508892
|
BRAND™
704504 |
5 to 25 mL | Non-sterile |
53.30 €
Pack of 3 |
In Stock
Log in to see stock. |
||||
10697045
|
BRAND™
704500 |
2 mL | Non-sterile |
36.35 €
Pack of 3 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
10693392
|
BRAND™
704508 |
25 mL | Sterile |
115.00 €
Pack of 5 |
In Stock
Log in to see stock. |
||||
10273201
|
BRAND™
704506 |
10 mL | Sterile |
110.00 €
Pack of 7 |
In Stock
Log in to see stock. |
||||
Description
- Sterile or non-sterile
- For use with seripettor™, seripettor™ pro and QuickSip bottletop dispensers
Specifications
Seripettor™ / Seripettor™ pro / QuikSip |