Learn More
Brady™ Brady™Bondz™ B-483 Ultra Aggressive Polyester BMP71 Printer Label
B-483 Ultra Aggressive Polyester Printer Labels are designed for use in commercial/industrial applications.
Brand:  Brady™ 115112
Product Code. 11701735
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Glossy white polyester with permanent rubber based adhesive is thermal transfer printable
  • Material is designed for high adhesion to textured metals and low surface energy plastics
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Service temperature of -40 to 248° F (-40 to 120° C)
Specifications
Specifications
UL Recognised; CSA Accepted
Permanent Rubber Based
Gloss
Non-sterile
1
Rectangle
0.004 in.
15.88 mm
25.4 mm
BMP71 Label Printer Labels
White
Polyester
BMP™71 Label Printer
500/Pk.
-40°C to +120°C
0.102 mm
0.5 x 1 in.
12.70 mm